Antibiotic Sensitivity Discs in Dispenser Vials
Supplier: LABORATORY MEDIA CORP SE
BB4331328
470102-494EA 35 USD
470102-494 470189-478
Antibiotic Sensitivity Discs in Dispenser Vials
Media for Microbiology Prepared Media for Microbiology
Each trigger-action magazine holds 50 discs, which are dispensed one at a time. These discs are excellent for qualitative testing of bacteria sensitivity. Measurements are in micrograms (except for penicillin).