Ward's® Live Bullfrogs (Rana catesbeiana)
Supplier: Avantor
0
470176-296PK 595.95 USD
470176-296 470180-614 470180-616 470189-248
Ward's® Live Bullfrogs (Rana catesbeiana)
Specimens Live Specimens Animals - Living Material
This large predatory frog will eat any other organism smaller than itself, including smaller bullfrogs; keep similar-sized frogs together to avoid cannibalism.
  • Large specimens
  • Excellent for observation in large aquaria.
This large predatory frog will eat any other organism smaller than itself, including smaller bullfrogs; keep similar-sized frogs together to avoid cannibalism. Frogs are 5" or larger in size.
Ordering Information: Contact your Sales Representative or call Customer Service to check availability.
Ordering information: These products contain living or perishable materials
This product is available to US customers only.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.