Pipe Stem Triangles
Supplier: United Scientific Supplies
CPT015 1350627020000
470189-154EA 3.1 USD
470189-154 470189-156
Pipe Stem Triangles
Tripods
Triangles made from galvanized iron wire, with porcelain pipe stemming to reduce wire sagging and oxidation.
  • Heavy gauge wire
  • Galvanized steel
Provides a patform to hold items over a Bunsen burner.