Esophagus Slide
Supplier: Avantor
470183-682EA 13.95 USD
470183-682
Esophagus Slide
Slides Prepared Slides
Extremely Thin Plastic Embedded Tissue For Greater Viewing Clarity
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Stratified squamous epithelium, glands of submucosa, and two layers of muscularis are clearly shown.