Zebra Fish Development Slide Set
Supplier: Avantor
470183-526KT 139.95 USD
470183-526
Zebra Fish Development Slide Set
Slides Prepared Slides
Illustrates Vertebrate Development
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Prepared in Ward's® own labs by skilled technicians, the slides show the detailed anatomy of organisms commonly used in genetic and developmental studies.
Ordering information: Set includes Cleavage stage (wm); Embryo stage (wm); Hatchling stage (wm); Adult female (ls); Adult male (ls); and Adult, composite (cs).