General Protist Survey Microscope Slide Set
Supplier: Avantor
470183-456KT 94.99 USD
470183-456
General Protist Survey Microscope Slide Set
Slides Prepared Slides
A Review of Sixteen Different Phyla
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
All are whole mounts unless otherwise specified.
Ordering information: Set includes Actinopod: Radiolaria strew; Archamoebae: Entamoeba histolytica cysts, Entamoeba coli trophozoites (sm), Archaezoa: Giardia trophozoites; Bacillariophyta (Stramenopila): Diatoms, mixed; Chlorophyta: Spirogyra, vegetative; Chrysophyta: Synura; Ciliata (Alveolata): Paramecium; Dinoflagellate: Peridinium; Euglenozoa: Euglena; Foraminfera: Foraminiferan strew; Myxomycota: Physarum plasmodium (sect); Oomycota: Saprolegnia; Phaeophyta (Kelp): Laminaria (sect); Rhodophyta: Batrachospermum; and Sarcodina (Rhizopod): Amoeba.