Plant Reproduction Slide Set
Supplier: Avantor
470183-446KT 145.99 USD
470183-446
Plant Reproduction Slide Set
Slides Prepared Slides
Examples of Reproductive Cells in Major Plant Divisions
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Ordering information: Set includes Bryophytes: Male Gamete: Marchantia, antheridia (ls); Female Gamete: Marchantia, archegonia (ls); Sporophyte: Marchantia, young sporophyte (mls); Zygote: Marchantia, mature sporophyte (ls); Ferns: Male Gamete: Fern prothallium, antheridia (sect); Female Gamete: Fern prothallium, archegonia (sect); Sporophyte: Fern prothallium, young sporophyte (wm); Fern, filamentous prothallium (wm); Gymnosperms: Male Gamete: Pinus, staminate cone (cs); Female Gamete: Pinus, ovulate cone (ls); Seed: Pinus, mature embryo (ls); Angiosperms: Flower: Lilium, young flower bud (ls); Male Gamete: Lilium, mature anther (cs); Female Gamete: Lilium ovary (cs); Seed: Lilium, mature seed with embryo (cs).