Barr Bodies, Male/Female Comparison Slide
Supplier: Avantor
470183-144EA 7.99 USD
470183-144
Barr Bodies, Male/Female Comparison Slide
Slides Prepared Slides
Illustrates Genetic "Femaleness" in Humans
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Scrapings from both human male and female mouth cavities on a single slide to demonstrate the presence of Barr bodies in the female cells and their absence in the male cells.