Barr Bodies Present Slide
Supplier: Avantor
470183-140EA 9.45 USD
470183-140
Barr Bodies Present Slide
Slides Prepared Slides
Illustrates Genetic "Femaleness" in Humans
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Smear of isolated squamous epithelial cells from the buccal cavity of a normal human female. Many nuclei show Barr bodies or drumstick chromosomes.