Peripheral Blood, Human Female Slide
Supplier: Avantor
470183-138EA 6.65 USD
470183-138
Peripheral Blood, Human Female Slide
Slides Prepared Slides
Note Female Blood Characteristics
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Normal. Barr bodies or drumsticks can be observed in at least 10% of the polymorphonuclear leucocytes.