Human Skin, Composite Slide
Supplier: Avantor
470183-070EA 12.2 USD
470183-070
Human Skin, Composite Slide
Slides Prepared Slides
Study human epidermis
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Pigmented and unpigmented adult skin with all layers, epidermis, dermis, and subcutaneous tissue, for easy comparison.