Parotid Gland, Human Slide
Supplier: Avantor
470183-040EA 6.65 USD
470183-040
Parotid Gland, Human Slide
Slides Prepared Slides
Major Salivary Gland
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Human. Shows serous salivary acini, ducts, interlobular connective tissue, and fat cells.