Entire Kidney Slide
Supplier: Avantor
470182-922EA 7.49 USD
470182-922
Entire Kidney Slide
Slides Prepared Slides
Observe Structures of the Kidney
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Rat. Through cortex, medulla, pelvis, and entrance of ureter.