Gallbladder, Mammal Slide
Supplier: Avantor
470182-888EA 9.95 USD
470182-888
Gallbladder, Mammal Slide
Slides Prepared Slides
Digestive Organ
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Through mucosa with simple columnar epithelium, serous layer, and muscular layer.