Small Intestine, Goblet Cells Slide
Supplier: Avantor
470182-874EA 7.45 USD
470182-874
Small Intestine, Goblet Cells Slide
Slides Prepared Slides
Investigate the digestive tract
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Specially stained to show the basement membrane, brush border of epithelium, and goblet cells.