Thyroid Gland, Mammal Slide
Supplier: Avantor
470182-792EA 8 USD
470182-792
Thyroid Gland, Mammal Slide
Slides Prepared Slides
Structures and components of the thyroid
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Shows hormone stored in follicles, as well as simple cuboidal cells, resting on reticular tissue, lining the follicles.