Medullated Nerve Fibers, Teased Preparation Slide
Supplier: Avantor
470182-690EA 17.99 USD
470182-690
Medullated Nerve Fibers, Teased Preparation Slide
Slides Prepared Slides
Investigate nerve structures
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Peripheral nerves treated with osmic acid. Excellent display of nodes of Ranvier and medullary sheaths.