Osteogenesis, Intramembranous Ossification Slide
Supplier: Avantor
470182-650EA 9.95 USD
470182-650
Osteogenesis, Intramembranous Ossification Slide
Slides Prepared Slides
Bone Development
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Demonstrates ossification of the skull and the flat bones of face.