Mast Cells Slide
Supplier: Avantor
470182-634EA 11.85 USD
470182-634
Mast Cells Slide
Slides Prepared Slides
Flat Mount of Mesentery
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Resorcin fuchsin stained to demonstrate mast cells as well as both white and elastic fibers.