Glycogen Slide
Supplier: Avantor
470182-594EA 8.5 USD
470182-594
Glycogen Slide
Slides Prepared Slides
Illustrates Energy Storage
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal liver. Glycogen granules stained red and nucleus blue. Compare to Glycogen Removed Slide.