Nucleic Acids Slide
Supplier: Avantor
470182-574EA 10.99 USD
470182-574
Nucleic Acids Slide
Slides Prepared Slides
Investigate Genetic Material Under the Microscope
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Onion root tip. DNA stains blue; RNA stains red.