Animal Meiosis, Spermatogenesis Slide
Supplier: Avantor
470182-560EA 12.5 USD
470182-560
Animal Meiosis, Spermatogenesis Slide
Slides Prepared Slides
Investigate spermatogenesis.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Grasshopper testis. Various stages of meiosis.