Fish and Onion Mitosis Slide
Supplier: Avantor
470182-556EA 29 USD
470182-556
Fish and Onion Mitosis Slide
Slides Prepared Slides
Compare and contrast mitosis
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Composite of three sections of fish blastodisc and three sections of onion root tip. Demonstrates animal and plant mitosis.