Menopon gallinae, Adult Slide
Supplier: Avantor
470182-236EA 11.5 USD
470182-236
Menopon gallinae, Adult Slide
Slides Prepared Slides
Ectoparasite
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Poultry louse. Unstained. Often found on the feather shaft of domestic chickens.