Entamoeba coli, Cysts Slide
Supplier: Avantor
470181-854EA 10.65 USD
470181-854
Entamoeba coli, Cysts Slide
Slides Prepared Slides
Intestinal Parasite
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
One to eight typical nuclei ; splinter shaped chromatoid bodies.