Amphioxus Slide
Supplier: Avantor
470181-686EA 14.95 USD
470181-686
Amphioxus Slide
Slides Prepared Slides
Explore early animal development stages
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Small adult exhibiting three main chordate characteristics: notochord, nerve chord, and pharyngeal clefts. It resembles both the echinoderms and vertebrates, providing a unique specimen for embryological study.