Honeybee Worker Leg, Composite Slide
Supplier: Avantor
470181-646EA 10.65 USD
470181-646
Honeybee Worker Leg, Composite Slide
Slides Prepared Slides
Illustrates Important Worker Bee Body Parts
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
One view each of the first, second, and third legs. Shows antenna cleaner, spur, and pollen basket. Unstained.