Springtail Slide
Supplier: Avantor
470181-608EA 5.49 USD
470181-608
Springtail Slide
Slides Prepared Slides
Usually Feed on Decaying Plant Matter
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Collembola. Shows the furcula, or jumping organ that characterizes this group. Unstained.