Pond Life, Microscrustacean Strew Slide
Supplier: Avantor
470181-602EA 14.75 USD
470181-602
Pond Life, Microscrustacean Strew Slide
Slides Prepared Slides
Illustrated the Varied Species within Aquatic Habitats
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mixture of the major microcrustacean types found in the open waters of ponds; includes copepods, cladocerans, ostracods, and branchiopods.