Ranunculus, Mature Root Slide
Supplier: Avantor
470181-380EA 6.95 USD
470181-380
Ranunculus, Mature Root Slide
Slides Prepared Slides
Illustrates characteristics of dicot structures
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Shows complete metaxylem. Clearly shows the compact vascular bundle of a dicot root.