Ward's® Sunflower (Helianthus) Slides
Supplier: Avantor
470177-466EA 6.5 USD
470177-466 470181-324 470181-326
Ward's® Sunflower (Helianthus) Slides
Slides Prepared Slides
A herbaceous dicot.
  • Ward's produced slides
  • Rigorous quality control standards
Helianthus is a great option for teaching about basic herbaceous dicot structure. The stems contain a very large pith, and the selection of young, mature, or combined sections makes it easy to see the growth of vascular bundles and increased organization as the stem ages.