Lily, Sporogenous Tissue
Supplier: Avantor
470181-246EA 6.99 USD
470181-246
Lily, Sporogenous Tissue
Slides Prepared Slides
Cross section, quadruple stained
  • All identifying features are clearly distinguishable
  • Rigorous quality control standards
Very young anther. Shows early cellular development prior to meiosis.