Lilium, Leaf Epidermis Slide
Supplier: Avantor
470181-242EA 7.75 USD
470181-242
Lilium, Leaf Epidermis Slide
Slides Prepared Slides
Illustrates Structure and Function of Gas Exchange
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Shows guard cells and stomata.