Acorus, Rhizome Slide
Supplier: Avantor
470181-224EA 6.55 USD
470181-224
Acorus, Rhizome Slide
Slides Prepared Slides
A typical hydrophyte.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Sweet flag. Shows amphivasal bundles and casparian strip.