Eudorina Slide
Supplier: Avantor
470180-974EA 5.25 USD
470180-974
Eudorina Slide
Slides Prepared Slides
Colonial Green algae
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Colonies of 16–64 cells; vegetative and reproductive stages.