Clostridium rubrum Slide
Supplier: Avantor
470180-894EA 8.25 USD
470180-894
Clostridium rubrum Slide
Slides Prepared Slides
Demonstrates Well-defined Endospore
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
An aerobic, spore forming, rod-shaped bacterium. Spores are oval in shape, and subterminal in location within the cell. This sometimes makes the cells appear to be ‘fatter’ at one end than the other.