Lambda DNA/EcoR I, Hind III Digest
Supplier: PROMEGA SUNNYVALE INC SE
G1731
470180-832EA 124.1 USD
470180-832
Lambda DNA/EcoR I, Hind III Digest
Electrophoresis Reagents Molecular Weight Markers
These high-quality DNA Markers were created by digesting specified DNA to completion with one or more restriction enzymes, complete with heat inactivation. The recommended loading for each is 1 {mu lower case}l per lane. Note: Perishable. Store frozen. Shipped next-day air on ice.
Supplied with 1 ml 6X blue/orange loading dye. 100μg (0.5μg/μL).