Benchtop DNA Marker
Supplier: PROMEGA SUNNYVALE INC SE
G8291
470180-822EA
318.35
USD
470180-822
Benchtop DNA Marker
Electrophoresis Reagents
Molecular Weight Markers
Ready to use, this DNA marker does not require preparation or refrigeration — it can be used effectively at room temperature. For additional convenience, the marker is supplied in blue/orange loading dye, so you can tract migration during electrophoresis. Load 6 μL per well. Total quantity: 300 μL.