Ward's® Prepared Yeast, Genetic (MIN) Agar
Supplier: IPM SCIENTIFIC, INC.
470180-814
470180-814PK 33.95 USD
470180-814
Ward's® Prepared Yeast, Genetic (MIN) Agar
Media for Microbiology Prepared Media for Microbiology
For Demonstration of Typtophan Dependance or Independence in Yeast
  • Sterile- no autoclaving
  • Convenient - prepared for you
  • Prepared consistenly every time
Ordering information: Some of these products contain living or perishable materials
Delivery information: Prepared plates will ship via overnight or 2nd day delivery.
Prepared bottles and tubes do not require as much temperature regulation in-transit so we ship them via ground to save you the cost of freight. During checkout, please specify a delivery date for your specimens to arrive.