Ward's® Prepared MacConkey Agar
Supplier: CULTURE MEDIA &SUPPLIES INC SE
CM102-P25
470180-742KT
31.75
USD
470180-742
Ward's® Prepared MacConkey Agar
Media for Microbiology
Prepared Media for Microbiology
For Differential Isolation of Enteric Bacilli Based on Lactose Fermentation
- Sterile- no autoclaving
- Convenient - prepared for you
- Prepared consistenly every time
Ordering information: These products contain living or perishable materials
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.