Ward's® Prepared Columbia CNA Agar
Supplier: IPM SCIENTIFIC, INC.
881702
470180-738PK
31.95
USD
470180-738
Ward's® Prepared Columbia CNA Agar
Media for Microbiology
Dehydrated Media for Microbiology
With 5% sheep’s blood. For isolation of Gram-positive organisms.
Ordering information: Sold only to colleges and accredited labs. Allow two weeks for delivery.
These products contain living or perishable materials
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.