Ward's® Prepared Brain-Heart Infusion Agar
Supplier: CULTURE MEDIA &SUPPLIES INC SE
CM115-P25
470180-706PK 35.95 USD
470180-706
Ward's® Prepared Brain-Heart Infusion Agar
Media for Microbiology Dehydrated Media for Microbiology
Brain-Heart Infusion (BHI) Agar is a general purpose media used for the cultivation of fastidous microrganisms.
Excellent media for cultivaion of such as streptococci, pneumococci and meningococci.
Ordering information: These products contain living or perishable materials
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.