Thioglycollate Medium
Supplier: Avantor
470180-662PK 41.95 USD
470180-662
Thioglycollate Medium
Media for Microbiology Dehydrated Media for Microbiology
Without indicator. For cultivation of anaerobic species and Clostridium. Suitable for fermentation studies where no indicator is required.