Nutrient Agar Deeps, Tubes
Supplier: Avantor
470180-660PK 34.99 USD
470180-660
Nutrient Agar Deeps, Tubes
Media for Microbiology Prepared Media for Microbiology
Contains 12 nutrient agar deeps. Used to check for organism motility. 17 mL per tube.