Sulfide Indole Motility (SIM) Agar Tubes
Supplier: Avantor
470180-658PK
29.95
USD
470180-658
Sulfide Indole Motility (SIM) Agar Tubes
Cell Culture Supplements and Additives
For the differentiation of members of the Enterobacteriacaea based on H2S production, indole production, and motility.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)