Halobacterium Agar
Supplier: Avantor
470180-654PK 43.95 USD
470180-654
Halobacterium Agar
Media for Microbiology Dehydrated Media for Microbiology
For cultivation of halophilic bacilli.
This set contains 2 tubes of Halobacterium Broth and 10 tubes of Halobacterium Agar.
Halophilic organisms like this one require a growth medium with a high salt content. As such, salt may be seen to precipitate out of the medium. This is not an indication of contamination or otherwise poor quality.