Triple Sugar Iron (TSI) Agar Tubes
Supplier: Avantor
470180-650PK 33.95 USD
470180-650
Triple Sugar Iron (TSI) Agar Tubes
Media for Microbiology Prepared Media for Microbiology
For identification of Gram-negative bacteria based on dextrose, lactose, and sucrose fermentation, and hydrogen sulfide production.