Mannitol Salt Agar
Supplier: CULTURE MEDIA &SUPPLIES INC SE
0
470180-646PK 24.99 USD
470180-646
Mannitol Salt Agar
Media for Microbiology Dehydrated Media for Microbiology
For cultivation of pathogenic staphylococci.