Chocolate Agar, Tubes
Supplier: CULTURE MEDIA &SUPPLIES INC SE
880088
470180-644PK 59.99 USD
470180-644
Chocolate Agar, Tubes
Media for Microbiology Dehydrated Media for Microbiology
Supplemented with X and V factors
For cultivation of Neisseria and Haemophilus species. Package of 12 tubes supplied.