Prepared Blood Agar Media
Supplier: CULTURE MEDIA &SUPPLIES INC SE
880070
880901
470180-642CS
354.99
USD
470180-642
470180-680
Prepared Blood Agar Media
Media for Microbiology
Dehydrated Media for Microbiology
Tryptic Soy Agar base with 5% Sheep Blood
- Excellent for cultivation of fastidious organisms
- Plate your organism onto Blood agar plates to test for hemolytic reaction
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)